Learn More
Tocris Bioscience™ 2-Phospho-L-ascorbic acid trisodium salt
GREENER_CHOICE
Ascorbic acid derivative; maintains differentiation potential in bone marrow-derived MSCs
Brand:  Tocris Bioscience™ 5778/50
Encompass
Product Code. 15707116
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
2-Phospho-L-ascorbic acid trisodium salt is a stable ascorbic acid derivative used in cell culture; in combination with FGF-2, maintains differentiation potential in bone marrow-derived mesenchymal stem cells (MSC) through increased expression of HGF. Also exhibits synergistic protection of hMSCs under oxidative stress in combination with N-acetylcysteine (Cat. No. 5619).
Specifications
Specifications
2-(Dihydrogen phosphate)-L-ascorbic acid sodium salt
66170-10-3