missing translation for 'onlineSavingsMsg'
Learn More

2-Phthalimidopropionic acid, 98%, Thermo Scientific™

Product Code. 11333127
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11333127 1 g 1g
11343127 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11333127

Brand: Thermo Scientific Alfa Aesar L13534.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19506-87-7
Molecular Formula C11H9NO4
Molecular Weight (g/mol) 219.196
MDL Number MFCD00023061
InChI Key OZWUITKBAWTEAQ-UHFFFAOYSA-N
Synonym 2-phthalimidopropionic acid, phthaloyl-dl-alanine, 2-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl propanoic acid, 2-1,3-dioxoisoindolin-2-yl propanoic acid, phthalyl-dl-alanine, 2-1,3-dioxoisoindol-2-yl propanoic acid, 2-1,3-dihydro-1,3-dioxo-2h-isoindol-2-yl propanoic acid, 2-phthalimidopropanoic acid, 2-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl propanoic acid, 2h-isoindole-2-acetic acid, 1,3-dihydro-alpha-methyl-1,3-dioxo
PubChem CID 86884
IUPAC Name 2-(1,3-dioxoisoindol-2-yl)propanoic acid
SMILES CC(C(=O)O)N1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 162°C to 164°C
Quantity 1 g
Beilstein 86452
Formula Weight 219.2
Percent Purity 98%
Chemical Name or Material 2-Phthalimidopropionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.