missing translation for 'onlineSavingsMsg'
Learn More

2-Picolinic acid, 99%

Product Code. 11467796
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11467796

Brand: Thermo Scientific Alfa Aesar A14749.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-98-6 | C6H5NO2 | 123.111 g/mol

2-Picolinic acid is used as a chelate for alkaline earth metals. Used to prepare picolinato ligated transition metal complexes. In synthetic organic chemistry, has been used as a substrate in the Mitsunobu reaction and in the Hammick reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Picolinic acid is used as a chelate for alkaline earth metals. Used to prepare picolinato ligated transition metal complexes. In synthetic organic chemistry, has been used as a substrate in the Mitsunobu reaction and in the Hammick reaction.

Solubility
Soluble in water. Also soluble in glacial acetic acid, ethanol and methanol. Insoluble in chloroform.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, reducing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-98-6
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.111
MDL Number MFCD00006293
InChI Key SIOXPEMLGUPBBT-UHFFFAOYSA-N
Synonym picolinic acid, 2-pyridinecarboxylic acid, 2-picolinic acid, 2-carboxypyridine, alpha-picolinic acid, o-pyridinecarboxylic acid, pyridinecarboxylic acid, alpha-pyridinecarboxylic acid, picolinate, acide picolique
PubChem CID 1018
ChEBI CHEBI:28747
IUPAC Name pyridine-2-carboxylic acid
SMILES C1=CC=NC(=C1)C(=O)O

Specifications

Melting Point 135°C to 139°C
Flash Point 139°C (232°F)
Odor Odorless
Quantity 100 g
Beilstein 109595
Merck Index 14,7403
Solubility Information Soluble in water. Also soluble in glacial acetic acid,ethanol and methanol. Insoluble in chloroform.
Formula Weight 123.11
Percent Purity 99%
Chemical Name or Material 2-Picolinic acid
compliance-icons compliance-icons
Product Identifier
  • 2-Picolinic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 2-Pyridinecarboxylicacid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.