missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Picolinic-d4 Acid, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 2-Picolinic-d4 Acid |
| CAS | 284487-61-2 |
| Molecular Formula | C6 D4 H N O2 |
| Quantity | 0.25 g |
| Synonym | 2-Pyridine-3,4,5,6-d4-carboxylic acid (9 CI), 2-Picolinic-d4 Acid, 2-Picolinic Acid-D4 |
| InChI Formula | InChI=1 S/C6H5NO2/c8-6(9)5-3-1-2-4-7-5/h1-4 H,(H,8,9)/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1nc(C(=O)O)c([2 H])c([2 H])c1[2 H] |
| IUPAC Name | 3,4,5,6-tetradeuteriopyridine-2-carboxylic acid |
| Molecular Weight (g/mol) | 127.134 |
| Formula Weight | 127.0571 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction