missing translation for 'onlineSavingsMsg'
Learn More

2-Picolyl chloride hydrochloride, 98%

Product Code. 10657342 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10657342 100 g Glass bottle 100g
10571811 5 g Glass bottle 5g
10255510 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10657342 Supplier Thermo Scientific Acros Supplier No. 172121000
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6959-47-3 | C6H6ClN·HCl | 164.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6959-47-3
Molecular Formula C6H6ClN·HCl
Molecular Weight (g/mol) 164.04
MDL Number MFCD00012811
InChI Key JPMRGPPMXHGKRO-UHFFFAOYSA-N
Synonym 2-chloromethyl pyridine hydrochloride, 2-picolyl chloride hydrochloride, 2-chloromethylpyridine hydrochloride, picolyl chloride hydrochloride, 2-chloromethyl pyridine hcl, 2-pyridylmethylchloride hydrochloride, 2-chloromethyl pyridinium chloride, pyridine, 2-chloromethyl-, hydrochloride, unii-yni4417kx0
PubChem CID 23392
ChEBI CHEBI:76600
IUPAC Name 2-(chloromethyl)pyridine;hydrochloride
SMILES C1=CC=NC(=C1)CCl.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 119.0°C to 125.0°C
Color Brown to Green-Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Formula Weight 164.04
Percent Purity 98%
Physical Form Crystalline Mass
Chemical Name or Material 2-Picolyl chloride hydrochloride, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Picolyl chloride hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.