missing translation for 'onlineSavingsMsg'
Learn More

2-Piperazin-1-ylnicotinic acid, Thermo Scientific™

Product Code. 10679110
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10679110 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10679110 Supplier Thermo Scientific Maybridge Supplier No. MAY00152DA
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 374063-94-2
Packaging Amber Glass Bottle
Molecular Formula C10H13N3O2
Quantity 1 g
Synonym 2-piperazin-1-ylnicotinic acid, 2-piperazin-1-yl pyridine-3-carboxylic acid, 3-pyridinecarboxylicacid, 2-1-piperazinyl, 2-piperazin-1-yl nicotinic acid, 2-piperazinylpyridine-3-carboxylic acid
InChI Key BCDHEUBISMKPOQ-UHFFFAOYSA-N
SMILES C1CN(CCN1)C2=C(C=CC=N2)C(=O)O
IUPAC Name 2-piperazin-1-ylpyridine-3-carboxylic acid
Molecular Weight (g/mol) 207.233
PubChem CID 2771623
Formula Weight 207.23
Percent Purity 95%
Chemical Name or Material 2-Piperazin-1-ylnicotinic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.