missing translation for 'onlineSavingsMsg'
Learn More

2-Piperidinemethanamine, TRC

Product Code. 30411966
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411966 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411966 Supplier Toronto Research Chemicals Supplier No. TRCP48196550G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (RS)-(piperidin-2-yl)methanamine
CAS 22990-77-8
Molecular Formula C6 H14 N2
Quantity 50 g
Synonym (RS)-(Piperidin-2-yl)methanamine, Flecainide Acetate Imp. B (EP)
InChI Formula InChI=1S/C6H14N2/c7-5-6-3-1-2-4-8-6/h6,8H,1-5,7H2
SMILES NCC1CCCCN1
IUPAC Name piperidin-2-ylmethanamine
Molecular Weight (g/mol) 114.19
Formula Weight 114.1157
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.