missing translation for 'onlineSavingsMsg'
Learn More

2-piperidinobenzamide, 97%, Thermo Scientific™

Product Code. 10604575
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10604575 25 g 25g
10026082 1 g 1g
10451913 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10604575 Supplier Thermo Scientific Maybridge Supplier No. BTB03853EB
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 3430-40-8
Molecular Formula C12H16N2O
Molecular Weight (g/mol) 204.27
MDL Number MFCD00052257
InChI Key VTXYPPVXMJMLCY-UHFFFAOYSA-N
Synonym 2-piperidinobenzamide, 2-piperidin-1-yl benzamide, benzamide,2-1-piperidinyl, 2-piperidylbenzamide, piperidinyl benzamide, maybridge1_000974, 2-1-piperidinyl benzamide, benzamide, 2-1-piperidinyl
PubChem CID 335067
SMILES NC(=O)C1=CC=CC=C1N1CCCCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 25 g
IUPAC Name 2-(piperidin-1-yl)benzamide
Formula Weight 204.27
Percent Purity 97%
Chemical Name or Material 2-Piperidinobenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.