missing translation for 'onlineSavingsMsg'
Learn More

2-Piperidinyl(1-pyrrolidinyl)methanone hydrochloride, ≥95%, Thermo Scientific™

Product Code. 10679135
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10679135 5 g 5g
10402082 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10679135 Supplier Thermo Scientific Maybridge Supplier No. MO01175DE
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 690634-81-2
Molecular Formula C10H19ClN2O
Molecular Weight (g/mol) 218.73
MDL Number MFCD06407992
InChI Key RIAUQORBEIQABE-UHFFFAOYNA-N
Synonym 2-pyrrolidine-1-carbonyl piperidine hydrochloride, 2-piperidinyl 1-pyrrolidinyl methanone hydrochloride, 2-pyrrolidin-1-yl carbonyl piperidine hydrochloride, piperidin-2-yl pyrrolidin-1-yl methanone hydrochloride, 1-2-piperidinylcarbonyl pyrrolidine hcl, 2-piperidyl pyrrolidinyl ketone, chloride, 2-pyrrolidin-1-ylcarbonyl piperidine hydrochloride
PubChem CID 2794693
IUPAC Name piperidin-2-yl(pyrrolidin-1-yl)methanone;hydrochloride
SMILES Cl.O=C(C1CCCCN1)N1CCCC1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Quantity 5 g
Formula Weight 218.73
Percent Purity 95%
Chemical Name or Material 2-Piperidinyl(1-pyrrolidinyl)methanone hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.