missing translation for 'onlineSavingsMsg'
Learn More

2-Propanesulfonyl chloride, 97%

Product Code. 11377705
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377705 5 g 5g
11387705 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11377705 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur L02431.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 10147-37-2 | C3H7ClO2S | 142.597 g/mol

2-Propanesulfonyl chloride is used as a pharmaceutical intermediate. Reductive cleavage of a secondary alcohol to methylene has been achieved by reduction of the 2-propanesulfonyl ester with lithium triethylborohydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Propanesulfonyl chloride is used as a pharmaceutical intermediate. Reductive cleavage of a secondary alcohol to methylene has been achieved by reduction of the 2-propanesulfonyl ester with lithium triethylborohydride.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases. Store under dry inert gas. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 10147-37-2
Molecular Formula C3H7ClO2S
Molecular Weight (g/mol) 142.597
MDL Number MFCD00007453
InChI Key DRINJBFRTLBHNF-UHFFFAOYSA-N
Synonym isopropylsulfonyl chloride, 2-propanesulfonyl chloride, isopropylsulphonyl chloride, isopropyl sulfonyl chloride, ccris 4139, 2-propanesulfonylchloride, isopropanesulfonyl chloride, iso-propylsulfonyl chloride, 2-propane sulfonyl chloride, isopropylsulphonyl choride
PubChem CID 82408
IUPAC Name propane-2-sulfonyl chloride
SMILES CC(C)S(=O)(=O)Cl

Spécification

Melting Point -47°C
Density 1.273
Boiling Point 95°C (45 mmHg)
Flash Point 86°C (186°F)
Refractive Index 1.456
Quantity 5 g
UN Number UN3265
Beilstein 1747497
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 142.6
Percent Purity 97%
Chemical Name or Material 2-Propanesulfonyl chloride
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.