missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, ACS Reagent, ≥99.5%, Honeywell Riedel-de Haën™

Product Code. 15651700
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 500 mL
20 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
1 L
2.5 L
20 L
500mL
Pack of 6
Product Code. Quantity Packaging unitSize
15651700 6 x 500 mL Glass Bottle Pack of 6
15631700 500 mL Glass Bottle 500mL
15601700 1 L Glass Bottle 1 L
15611700 2.5 L Glass Bottle 2.5 L
15641700 6 x 1 L Glass Bottle Pack of 6
15621700 20 L Stainless Steel Can 20 L
6 options
This item is not returnable. View return policy

Product Code. 15651700

Brand: Honeywell-Riedel-de Haen 1907646X500ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ACS reagent,≥99.5%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -90°C
Color Colorless
Density 0.785g/mL at 25°C
Boiling Point 83°C
Quantity 6 x 500 mL
Explosion Limit 0.02
Flash Point 12°C (53.6°F)
Ignition Point 750°F
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.377(lit.)
Linear Formula (CH3)2CHOH
Vapor Pressure 44mmHg (25°C), 33mmHg (20°C)
UN Number UN1219
Beilstein 635639
Solubility Information Soluble in water
Formula Weight 60.1g/mol
Percent Purity ≥99.5%
Grade ACS Reagent
Chemical Name or Material 2-Propanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.