missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, Puriss. p.a., ACS Reagent, ≥99.8% (GC), Honeywell Riedel-de Haën™

Product Code. 15665890
Click to view available options
Quantity:
4 x 25 mL
4 x 2.5 L
6 x 1 L
155 kg
Packaging:
Glass Bottle
Metal Drum
Unit Size:
155kg
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 15665890

Brand: Honeywell-Riedel-de Haen 593004X25ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent,≥99.8% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -89.5°C (literature)
CAS Max % 100.0000%
Density 0.785 g/mL at 20 °C; 0.785 g/mL at 25 °C(lit.)
Boiling Point 77°C to 83°C
Quantity 4 x 25 mL
Explosion Limit 0.02
Flash Point 12°C
Packaging Glass Bottle
Refractive Index 1.378
Linear Formula (CH3)2CHOH
Vapor Pressure 44mmHg (25°C), 33mmHg (20°C)
UN Number UN1219
Viscosity 4.3 mPas at 20°C
Beilstein 635639
Molecular Weight (g/mol) 60.10
Formula Weight 60.11
Percent Purity >99.8%
Grade puriss. p.a.
Chemical Name or Material 2-Propanol, Puriss. p.a., ACS Reagent, ≥99.8% (GC)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.