missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, puriss. p.a., Reag. ACS, Reag. ISO, Reag. PH. EUR., Reag. USP, Honeywell Riedel-de Haën™

Product Code. 15686890
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Plastic Bottle
Poly Bottle
Unit Size:
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 15686890

Brand: Honeywell-Riedel-de Haen 722992.5L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

puriss. p.a., Reag. ACS, Reag. ISO, Reag. PH. EUR., Reag. USP

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

CAS Max % 100.0000%
Density 0.784 - 0.786 g/cm3
Boiling Point 77°C to 83°C
Quantity 2.5 L
Flash Point 12°C
Packaging Poly Bottle
UN Number UN1219
Beilstein 635639
Formula Weight 60.11
Percent Purity >99.9%
Grade ACS Reagent
Chemical Name or Material 2-Propanol, puriss. p.a., Reag. ACS, Reag. ISO, Reag. PH. EUR., Reag. USP
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Propanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.