missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, Spectrophotometric Grade, 99.7+%

Product Code. 11343578
Change view
Click to view available options
Quantity:
1 L
4 x 1 L
Unit Size:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343578 1 L 1 L
11323578 4 x 1 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343578 Supplier Thermo Scientific Alfa Aesar Supplier No. 039194.K2
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 67-63-0 | C3H8O | 60.096 g/mol

Solvent for gums, resins, alkaloids, essential oils. Denaturant for ethyl alcohol. In antifreeze compositions. It is used in lotions and cosmetics. Isopropyl alcohol is used in keyboard, LCD and laptop cleaning. It is used as a wetting agent in the fountain solution used in lithographic printing, and often used as a solvent for French polishing shellac used in cabinet making. It act as hydride source in the Meerwein-Ponndorf-Verley reduction

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.096
MDL Number MFCD00011674
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O

Specifications

Melting Point -90°C
Density 0.785 g/mL
Boiling Point 82°C to 83°C
Quantity 1 L
Flash Point 12°C (53°F)
Odor Alcohol-like
Physical Form Liquid
Refractive Index 1.377
UN Number UN1219
Beilstein 635639
Merck Index 14,5208
Solubility Information Miscible with water,ethanol,acetone,chloroform,methanol,diethyl ether,n-octanol and benzene.
Formula Weight 60.1
Percent Purity ≥99.7%
Grade Spectrophotometric
Chemical Name or Material 2-Propanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.