missing translation for 'onlineSavingsMsg'
Learn More

2-(Propylthio)nicotinic acid, 97%, Thermo Scientific™

Product Code. 10744573
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10744573 1 g Amber Glass Bottle 1g
10455302 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Artikelnummer. 10744573 Leverandør Thermo Scientific Maybridge Leverandørnr. BTB06368DA
 Restricted Item
Estimated Shipment: 10-11-2026
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Kemiske identifikatorer

CAS 175135-22-5
Molecular Formula C9H10NO2S
Molecular Weight (g/mol) 196.24
MDL Number MFCD00051682
InChI Key CSMDLVRDBYKTAH-UHFFFAOYSA-M
Synonym 2-propylthio nicotinic acid, 2-n-propylthio nicotinic acid, 2-propylsulfanyl pyridine-3-carboxylic acid, 3-pyridinecarboxylicacid, 2-propylthio, 2-propylthiopyridine-3-carboxylic acid, maybridge1_001579, 2-prop-1-ylsulphanyl pyridine-3-carboxylic acid, 2-propylsulfanyl-nicotinic acid
PubChem CID 2774526
IUPAC Name 2-propylsulfanylpyridine-3-carboxylic acid
SMILES CCCSC1=NC=CC=C1C([O-])=O

Tekniske data

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 197.26
Percent Purity 97%
Chemical Name or Material 2-(Propylthio)nicotinic acid
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.