missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrid-4-ylpyrimidine-5-carboxylic acid, 90%, Thermo Scientific™

Product Code. 10346764
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Packaging Dimensione della confezione
10346764 250 mg Amber Glass Bottle 250mg
10022354 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 10346764 Fornitore Thermo Scientific Maybridge N. del fornitore CC66601CB
 Restricted Item
Estimated Shipment: 19-01-2027
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Identificatori chimici

CAS 259807-47-1
Molecular Formula C10H7N3O2
Molecular Weight (g/mol) 201.185
MDL Number MFCD09863218
InChI Key GTKJGOOMSODGOU-UHFFFAOYSA-N
Synonym 2-pyridin-4-yl pyrimidine-5-carboxylic acid, 2-pyrid-4-ylpyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 2-4-pyridinyl, 2-4-pyridyl pyrimidine-5-carboxylic acid, 2-4-pyridinyl-5-pyrimidinecarboxylic acid, 2-pyridin-4-yl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylic acid, 2-4-pyridinyl, 5-pyrimidinecarboxylic acid, 2-4-pyridinyl-9ci
PubChem CID 22710213
IUPAC Name 2-pyridin-4-ylpyrimidine-5-carboxylic acid
SMILES C1=CN=CC=C1C2=NC=C(C=N2)C(=O)O

Specifica

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 201.18
Percent Purity 90%
Chemical Name or Material 2-Pyrid-4-ylpyrimidine-5-carboxylic acid
Titolo del prodotto
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.