missing translation for 'onlineSavingsMsg'
Learn More

2-Pyridylacetic acid hydrochloride, 99%

Product Code. 10336970 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10336970 5 g Glass bottle 5g
10274800 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10336970 Supplier Thermo Scientific Acros Supplier No. 131950050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 16179-97-8 | C7H7NO2·HCl | 173.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16179-97-8
Molecular Formula C7H7NO2·HCl
Molecular Weight (g/mol) 173.6
MDL Number MFCD00012812
InChI Key MQVISALTZUNQSK-UHFFFAOYSA-N
Synonym 2-pyridylacetic acid hydrochloride, 2-pyridin-2-yl acetic acid hydrochloride, 2-pyridineacetic acid hydrochloride, 2-pyridineacetic acid, hydrochloride, pyridin-2-ylacetic acid hydrochloride, pyridylacetic acid hydrochloride, 2-pyridineacetic acid, hydrochloride 1:1, 2-pyridylacetic acid hydrochloide, pyridin-2-yl acetic acid hydrochloride, 2-2-pyridyl acetic acid hydrochloride
PubChem CID 85317
IUPAC Name 2-pyridin-2-ylacetic acid;hydrochloride
SMILES C1=CC=NC(=C1)CC(=O)O.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 135°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (C5H4N)CH2COOH·HCl
Quantity 5 g
Formula Weight 173.6
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Pyridylacetic acid hydrochloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Pyridylacetic acid hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.