missing translation for 'onlineSavingsMsg'
Learn More

2-pyrrolidin-1-ylaniline, Thermo Scientific™

Product Code. 10777475
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10777475 5 g 5g
10295784 250 mg 250mg
10091284 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10777475 Supplier Thermo Scientific Maybridge Supplier No. CC44814DE
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 21627-58-7
Molecular Formula C10H14N2
Molecular Weight (g/mol) 162.236
InChI Key BFPUBGCFJMIZDF-UHFFFAOYSA-N
Synonym 2-pyrrolidin-1-yl aniline, 2-pyrrolidinoaniline, 2-pyrrolidin-1-yl-phenylamine, 2-pyrrolidinylphenylamine, 2-pyrrolizinoaniline, acmc-209fmh, 2-pyrrolidin-1-ylphenylamine, 1-2-aminophenyl pyrrolidine, 2-pyrrolidin-1-ylphenyl amine, 2-1-pyrrolidinyl-benzenamine
PubChem CID 937501
IUPAC Name 2-pyrrolidin-1-ylaniline
SMILES C1CCN(C1)C2=CC=CC=C2N

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 162.23
Percent Purity 97%
Chemical Name or Material 2-Pyrrolidin-1-ylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.