missing translation for 'onlineSavingsMsg'
Learn More

2-Pyrrolidinone, 99%

Product Code. 11419150
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419150 2 kg 2kg
11429150 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419150 Supplier Thermo Scientific Alfa Aesar Supplier No. A15332.A3
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 616-45-5 | C4H7NO | 85.106 g/mol

2-Pyrrolidinone is used as a derivative in pharmaceuticals such as cotinine, doxapram, piracetam, povidone and ethosuximide. It is used as a solvent and an intermediate in polymers, inkjet cartridges, tone and colorant products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Pyrrolidinone is used as a derivative in pharmaceuticals such as cotinine, doxapram, piracetam, povidone and ethosuximide. It is used as a solvent and an intermediate in polymers, inkjet cartridges, tone and colorant products.

Solubility
Miscible with water and other organic solvents.

Notes
Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-45-5
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.106
MDL Number MFCD00005270
InChI Key HNJBEVLQSNELDL-UHFFFAOYSA-N
Synonym 2-pyrrolidinone, pyrrolidone, butyrolactam, 2-pyrrolidone, 2-oxopyrrolidine, pyrrolidon, 2-ketopyrrolidine, 2-pyrol, pyrrolidinone, gamma-butyrolactam
PubChem CID 12025
ChEBI CHEBI:36592
IUPAC Name pyrrolidin-2-one
SMILES C1CC(=O)NC1

Specifications

Melting Point 23°C to 25°C
Density 1.12
Boiling Point 245°C
Flash Point >110°C (230°F)
Refractive Index 1.4875
Quantity 2 kg
Sensitivity Hygroscopic
Merck Index 14,8016
Solubility Information Miscible with water and other organic solvents.
Formula Weight 85.11
Percent Purity 99%
Chemical Name or Material 2-Pyrrolidinone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Pyrrolidinone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H319-Causes serious eye irritation.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-Pyrrolidinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.