missing translation for 'onlineSavingsMsg'
Learn More

2-Quinoxalinecarbaldehyde, 97+%, Thermo Scientific™

Product Code. 10739783
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10739783 250 mg Amber Glass Bottle 250mg
10676753 1 g Amber Glass Bottle 1g
10547464 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10739783 Supplier Thermo Scientific Maybridge Supplier No. CC03504CB
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1593-08-4
Molecular Formula C9H6N2O
Molecular Weight (g/mol) 158.16
MDL Number MFCD00272280
InChI Key UJEHWLFUEQHEEZ-UHFFFAOYSA-N
Synonym 2-formylquinoxaline, 2-quinoxalinecarbaldehyde, 2-quinoxalinecarboxaldehyde, quinoxaline-2-carboxaldehyde, chinoxalin-2-aldehyd, pubchem21935, rarechem ak ml 0126, art-chem-bb b000386, quinoxalin-2-carboxaldehyde, 2-quinoxaline-carboxaldehyde
PubChem CID 594088
IUPAC Name quinoxaline-2-carbaldehyde
SMILES O=CC1=CN=C2C=CC=CC2=N1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 158.16
Percent Purity 97%
Chemical Name or Material 2-Quinoxalinecarbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.