missing translation for 'onlineSavingsMsg'
Learn More

2-Quinoxalinecarboxaldehyde, TRC

Product Code. 30409732
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409732 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409732 Supplier Toronto Research Chemicals Supplier No. TRCQ765200250MG
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1593-08-4
Molecular Formula C9H6N2O
Molecular Weight (g/mol) 158.16
Synonym 2-Formylquinoxaline, Quinoxalin-2-carboxaldehyde
IUPAC Name quinoxaline-2-carbaldehyde
SMILES O=Cc1cnc2ccccc2n1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H6N2O/c12-6-7-5-10-8-3-1-2-4-9(8)11-7/h1-6H
Formula Weight 158.048
Chemical Name or Material 2-Quinoxalinecarboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.