missing translation for 'onlineSavingsMsg'
Learn More

2-Quinoxalinecarboxylic Acid, TRC

Product Code. 30134255
Change view
Click to view available options
Quantity:
500 mg
1 g
10 g
Unit Size:
10g
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134255 10 g 10g
30004204 500 mg 500mg
30006650 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134255 Supplier Toronto Research Chemicals Supplier No. TRCQ76526510G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 879-65-2
Molecular Formula C9 H6 N2 O2
Molecular Weight (g/mol) 174.16
Synonym Quinoxaline-2-carboxylic acid, 2-Carboxyquinoxaline
IUPAC Name quinoxaline-2-carboxylic acid
SMILES OC(=O)c1cnc2ccccc2n1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H6N2O2/c12-9(13)8-5-10-6-3-1-2-4-7(6)11-8/h1-5H,(H,12,13)
Formula Weight 174.0429
Chemical Name or Material 2-Quinoxalinecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.