missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butoxy-6-chloropyridine, 97%

Product Code. 15461228
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15461228 1 g 1g
15471228 5 g 5g
15481228 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15461228 Supplier Thermo Scientific Alfa Aesar Supplier No. H55085.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 547740-43-2 | C9H12ClNO | 185.651 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 547740-43-2
Molecular Formula C9H12ClNO
Molecular Weight (g/mol) 185.651
MDL Number MFCD08705263
InChI Key PLDRTFJRLUKTTA-UHFFFAOYSA-N
Synonym 2-tert-butoxy-6-chloropyridine, acmc-20anmt, 2-tert-butoxy-6-chloro-pyridine, pyridine, 2-chloro-6-1,1-dimethylethoxy
PubChem CID 11984350
IUPAC Name 2-chloro-6-[(2-methylpropan-2-yl)oxy]pyridine
SMILES CC(C)(C)OC1=NC(=CC=C1)Cl

Specifications

Color Colorless
Density 1.076 g/mL
Flash Point 78°C (172°F)
Quantity 1 g
Formula Weight 185.65
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-tert-Butoxy-6-chloropyridine

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.