missing translation for 'onlineSavingsMsg'
Learn More

[2-(tert-Butoxy)ethyl](methyl)amine, TRC

Product Code. 30329163
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30329163 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329163 Supplier Toronto Research Chemicals Supplier No. TRCB416463500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material [2-(tert-Butoxy)ethyl](methyl)amine
CAS 1008119-69-4
Molecular Formula C7H17NO
Quantity 500 mg
InChI Formula InChI=1S/C7H17NO/c1-7(2,3)9-6-5-8-4/h8H,5-6H2,1-4H3
SMILES CNCCOC(C)(C)C
IUPAC Name N-methyl-2-[(2-methylpropan-2-yl)oxy]ethanamine
Molecular Weight (g/mol) 131.22
Formula Weight 131.131
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.