missing translation for 'onlineSavingsMsg'
Learn More

2-[(tert-Butoxycarbonyl)amino]-3-(3-fluorophenyl)propanoic acid, Tech., Thermo Scientific™

Product Code. 10643255
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10643255

Brand: Thermo Scientific Maybridge SB02000DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 114873-01-7
Molecular Formula C14H18FNO4
Molecular Weight (g/mol) 283.299
MDL Number MFCD00672522
InChI Key FPCCREICRYPTTL-NSHDSACASA-N
Synonym boc-phe 3-f-oh, boc-l-3-fluorophenylalanine, boc-3-fluoro-l-phenylalanine, boc-l-phe 3-f-oh, s-2-tert-butoxycarbonyl amino-3-3-fluorophenyl propanoic acid, boc-l-3-fluorophe, boc-3-fluoro-l-phe, 2s-2-tert-butoxy carbonyl amino-3-3-fluorophenyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-3-fluorophenyl propanoic acid, n-boc-3-fluoro-l-phenylalanine
PubChem CID 2734492
IUPAC Name (2S)-3-(3-fluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC(=CC=C1)F)C(=O)O

Specifications

Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 283.3
Percent Purity Tech.
Grade Technical
Chemical Name or Material (S)-N-(tert-Butoxycarbonyl)-3-fluorophenylalanine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.