missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butyl-4-methylthiazole-5-methanol, 97%, Thermo Scientific™

Product Code. 15445047
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15445047 5 g 5g
15455047 250 mg 250mg
15435047 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15445047 Supplier Thermo Scientific Alfa Aesar Supplier No. H32329.06
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1256483-61-0
Molecular Formula C9H15NOS
Molecular Weight (g/mol) 185.29
MDL Number MFCD16660039
InChI Key HJESPMQKUUFUGH-UHFFFAOYSA-N
Synonym 2-tert-butyl-4-methylthiazole-5-methanol, 2-tert-butyl-4-methyl-1,3-thiazol-5-yl methanol, 2-tert-butyl-5-hydroxymethyl-4-methylthiazole
PubChem CID 73012579
IUPAC Name (2-tert-butyl-4-methyl-1,3-thiazol-5-yl)methanol
SMILES CC1=C(CO)SC(=N1)C(C)(C)C

Specifications

Density 1.085
Boiling Point 110°C (0.75 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.525
Quantity 5 g
Formula Weight 185.29
Percent Purity 97%
Chemical Name or Material 2-tert-Butyl-4-methylthiazole-5-methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.