missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butyl-trans-vinylboronic acid, 97%

Product Code. 15499377
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15499377 250 mg 250mg
15489377 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15499377 Supplier Thermo Scientific Alfa Aesar Supplier No. H52627.MD
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 86595-37-1 | C6H13BO2 | 127.978 g/mol

2-tert-Butyl-trans-vinylboronic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-tert-Butyl-trans-vinylboronic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86595-37-1
Molecular Formula C6H13BO2
Molecular Weight (g/mol) 127.978
MDL Number MFCD01074624
InChI Key CTIWKIMYFQSVBZ-SNAWJCMRSA-N
Synonym 3,3-dimethyl-1-butenylboronic acid, e-3,3-dimethylbut-1-en-1-yl boronic acid, 2-t-butyl-e-vinylboronic acid, 2-tert-butyl-e-vinylboronic acid, e-3,3-dimethylbut-1-enyl boronic acid, 1-butenyl-3,3-dimethyboronic acid, 1e-3,3-dimethylbut-1-en-1-ylboronic acid, boronic acid, b-3,3-dimethyl-1-buten-1-yl, 3,3-dimethylbut-1-enylboronic acid
PubChem CID 5706775
IUPAC Name [(E)-3,3-dimethylbut-1-enyl]boronic acid
SMILES B(C=CC(C)(C)C)(O)O

Specifications

Melting Point 78°C to 80°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 127.98
Percent Purity 97%
Chemical Name or Material 2-tert-Butyl-trans-vinylboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.