missing translation for 'onlineSavingsMsg'
Learn More

2-(tert-butylsulfonyl)-3-ethoxyacrylonitrile, Thermo Scientific™

Product Code. 10368322
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10368322 1 g 1g
10739923 10 g 10g
10600286 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10368322 Supplier Thermo Scientific Maybridge Supplier No. KM01862DA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 175201-67-9
Molecular Formula C9H15NO3S
Molecular Weight (g/mol) 217.283
InChI Key UEZQDEIGENVNHG-FPLPWBNLSA-N
Synonym 2z-3-ethoxy-2-2-methylpropane-2-sulfonyl prop-2-enenitrile
PubChem CID 40455567
IUPAC Name (Z)-2-tert-butylsulfonyl-3-ethoxyprop-2-enenitrile
SMILES CCOC=C(C#N)S(=O)(=O)C(C)(C)C

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 217.29
Percent Purity 97%
Chemical Name or Material 2-(tert-Butylsulfonyl)-3-ethoxyacrylonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.