missing translation for 'onlineSavingsMsg'
Learn More

2-Tetrahydropyran-4-ylethanol, 97%, Thermo Scientific™

Product Code. 10792164
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10792164 1 g 1g
10472074 250 mg 250mg
10612315 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10792164 Supplier Thermo Scientific Maybridge Supplier No. CC67409DA
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 4677-18-3
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00129068
InChI Key XZXZZACRGBBWTQ-UHFFFAOYSA-N
Synonym 2-tetrahydro-2h-pyran-4-yl ethanol, 2-oxan-4-yl ethan-1-ol, 2-tetrahydro-pyran-4-yl-ethanol, 2-tetrahydropyran-4-ylethanol, 2-oxan-4-yl ethanol, 4-2-hydroxyethyl oxane, 4-hydroxyethyl tetrahydropyran, 2h-pyran-4-ethanol, tetrahydro, 2-tetrahydro-2h-pyran-4-yl ethan-1-ol, 2-tetrahydro-2h-pyran-4-ylethanol
PubChem CID 17750944
SMILES OCCC1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 2-(oxan-4-yl)ethanol
Formula Weight 130.19
Percent Purity 97%
Chemical Name or Material 2-Tetrahydropyran-4-ylethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.