missing translation for 'onlineSavingsMsg'
Learn More

2-(Tetrazol-5-yl)phenylboronic acid, 95%

Product Code. 11996751 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11996751

Brand: Thermo Scientific Acros 445550010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 155884-01-8 | C7H7BN4O2 | 189.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155884-01-8
Molecular Formula C7H7BN4O2
Molecular Weight (g/mol) 189.97
MDL Number MFCD06796325
InChI Key GVRXWYFECKHTSJ-UHFFFAOYSA-N
Synonym 2-5-tetrazolyl phenylboronic acid, 2-1h-tetrazol-5-yl phenyl boronic acid, 2-tetrazol-5-yl phenylboronic acid, 2-2h-tetrazol-5-yl phenylboronic acid, 2-2h-tetrazol-5-yl-phenylboronic acid, 2-1h-tetrazol-5-yl phenylboronic acid, 2-2h-tetrazol-5-yl phenyl boronic acid, boronic acid, 2-1h-tetrazol-5-yl phenyl, boronic acid,b-2-2h-tetrazol-5-yl phenyl
PubChem CID 11171482
IUPAC Name [2-(2H-1,2,3,4-tetrazol-5-yl)phenyl]boronic acid
SMILES OB(O)C1=CC=CC=C1C1=NNN=N1

Specifications

Melting Point 148°C to 152°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 189.97
Percent Purity 95%
Chemical Name or Material 2-(Tetrazol-5-yl)phenylboronic acid
compliance-icons
Product Identifier
  • 2-(Tetrazol-5-yl)phenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.