missing translation for 'onlineSavingsMsg'
Learn More

2-Thiazolecarboxaldehyde, 98%

Product Code. 10534793 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10534793 1 g Glass bottle 1g
11976771 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10534793 Supplier Thermo Scientific Acros Supplier No. 377680010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10200-59-6 | C4H3NOS | 113.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10200-59-6
InChI Key ZGTFNNUASMWGTM-UHFFFAOYSA-N
Synonym 2-thiazolecarboxaldehyde, thiazole-2-carbaldehyde, 2-formylthiazole, thiazole-2-carboxaldehyde, 2-formyl thiazole, thiazolecarboxaldehyde, 2-formyl-1,3-thiazole, 1,3-thiazole-2-carboxaldehyde, 2-thiazolecarbaldehyde, pubchem9929
PubChem CID 2734903
ChEBI CHEBI:43623
IUPAC Name 1,3-thiazole-2-carbaldehyde
SMILES C1=CSC(=N1)C=O

Specifications

Color Yellow to Brown
Density 1.2880g/mL
Boiling Point 76.0°C to 78.0°C (24.0 mmHg)
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C4H3NOS
Refractive Index 1.5735 to 1.5755
Quantity 1 g
Specific Gravity 1.288
Solubility Information Solubility in water: soluble.
Molecular Weight (g/mol) 113.14
Formula Weight 113.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Thiazolecarboxaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-Thiazolecarboxaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.