missing translation for 'onlineSavingsMsg'
Learn More

2-Thien-2-ylpyrimidine-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10081574
Change view
Click to view available options
:
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10081574 Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10081574 Supplier Thermo Scientific Maybridge Supplier No. CC66501DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 916766-97-7
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C9H6N2O2S
MDL Number MFCD09817551
Quantity 1 g
Synonym 2-thien-2-ylpyrimidine-5-carboxylic acid, 2-thiophen-2-yl pyrimidine-5-carboxylic acid, 2-2-thienyl-5-pyrimidinecarboxylic acid, 2-2-thienyl pyrimidine-5-carboxylic acid, 2-thien-2-yl pyrimidine-5-carboxylic acid
InChI Key SJJLOOUOVNRKLF-UHFFFAOYSA-N
SMILES C1=CSC(=C1)C2=NC=C(C=N2)C(=O)O
IUPAC Name 2-thiophen-2-ylpyrimidine-5-carboxylic acid
Molecular Weight (g/mol) 206.219
PubChem CID 24229728
Formula Weight 206.22
Percent Purity 97%
Chemical Name or Material 2-Thien-2-ylpyrimidine-5-carboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.