missing translation for 'onlineSavingsMsg'
Learn More

2-Thienylmagnesium bromide, 1M solution in THF, AcroSeal™

Product Code. 10496709 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
50mL
This item is not returnable. View return policy

Product Code. 10496709

Brand: Thermo Scientific Acros 434610500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5713-61-1 | C4H3BrMgS | 187.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 65.0°C
CAS 5713-61-1, 109-99-9
Color Brown to Yellow
CAS Min % 17.6, 79.62
CAS Max % 20.38, 82.4
Physical Form Solution
Quantity 50 mL
Molecular Formula C4H3BrMgS
MDL Number MFCD03094046
Flash Point −21°C
Synonym 2-thienylmagnesium bromide, thiophen-2-yl-magnesium bromide 1.0m, 2-thienylmagnesium bromide solution, thienylmagnesiumbromide, thienylmagnesium bromide, thienyl magnesium bromide, 2-thienyl magnesium bromide, bromo thiophen-2-yl magnesium, thiophen-2-ylmagnesium bromide, thiphen-2-yl magnesium bromide, grignard reagent
Solubility Information Solubility in water: reacts
InChI Key GSHPYJFNTAMRJF-UHFFFAOYSA-M
SMILES Br[Mg]C1=CC=CS1
IUPAC Name magnesium;2H-thiophen-2-ide;bromide
Molecular Weight (g/mol) 187.34
PubChem CID 11074151
Formula Weight 187.34
Concentration or Composition (by Analyte or Components) 0.95 to 1.10M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.011
Density 1.0110g/mL
Chemical Name or Material 2-Thienylmagnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Thienylmagnesium bromide, 1M solution in THF
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.