missing translation for 'onlineSavingsMsg'
Learn More

2-Thiomorpholinoisonicotinonitrile, 97%, Thermo Scientific™

Product Code. 10327444
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10327444 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10327444 Supplier Thermo Scientific Maybridge Supplier No. CC50516DA
 Restricted Item
Estimated Shipment: 08-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 884507-30-6
Molecular Formula C10H11N3S
Molecular Weight (g/mol) 205.28
MDL Number MFCD09025862
InChI Key MZIANIKUVGCEPB-UHFFFAOYSA-N
Synonym 2-thiomorpholinoisonicotinonitrile, 2-thiomorpholin-4-yl pyridine-4-carbonitrile, 4-pyridinecarbonitrile,2-4-thiomorpholinyl, 2-1,4-thiazaperhydroin-4-yl pyridine-4-carbonitrile
PubChem CID 18525850
IUPAC Name 2-(thiomorpholin-4-yl)pyridine-4-carbonitrile
SMILES N#CC1=CC(=NC=C1)N1CCSCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 205.28
Percent Purity 97%
Chemical Name or Material 2-Thiomorpholinoisonicotinonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.