missing translation for 'onlineSavingsMsg'
Learn More

(2-Thiomorpholinopyrid-4-yl)methanol, 97%, Thermo Scientific™

Product Code. 10193464
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10193464 250 mg Amber Glass Bottle 250mg
10091824 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10193464 Supplier Thermo Scientific Maybridge Supplier No. CC50509CB
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 898289-25-3
Molecular Formula C10H14N2OS
Molecular Weight (g/mol) 210.295
MDL Number MFCD09702362
InChI Key KDJBEJVGUFHDPW-UHFFFAOYSA-N
Synonym 2-thiomorpholinopyrid-4-yl methanol, 2-thiomorpholin-4-yl pyridin-4-yl methanol, 2-thiomorpholin-4-ylpyridin-4-yl methanol, 4-pyridinemethanol,2-4-thiomorpholinyl, 4-hydroxymethyl-2-thiomorpholin-4-yl pyridine, 2-thiomorpholinopyridin-4-yl methanol, 4-4-hydroxymethyl pyridin-2-yl thiomorpholine, 2-1,4-thiazaperhydroin-4-yl-4-pyridyl methan-1-ol
PubChem CID 24229508
IUPAC Name (2-thiomorpholin-4-ylpyridin-4-yl)methanol
SMILES C1CSCCN1C2=NC=CC(=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 210.3
Percent Purity 97%
Chemical Name or Material (2-Thiomorpholinopyrid-4-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.