missing translation for 'onlineSavingsMsg'
Learn More

(2-Thiomorpholinopyrid-4-yl)methylamine, 97%, Thermo Scientific™

Product Code. 10752514
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10752514 250 mg 250mg
10693055 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10752514 Supplier Thermo Scientific Maybridge Supplier No. CC50513CB
 Restricted Item
Estimated Shipment: 08-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 886851-35-0
Molecular Formula C10H15N3S
Molecular Weight (g/mol) 209.311
MDL Number MFCD09025861
InChI Key FFHDQWVSXSKWLF-UHFFFAOYSA-N
Synonym 2-thiomorpholinopyrid-4-yl methylamine, 2-thiomorpholin-4-yl pyridin-4-yl methanamine, 2-thiomorpholinopyridin-4-yl methanamine, 1-2-thiomorpholin-4-yl pyridin-4-yl methanamine, 2-thiomorpholinopyridin-4-yl methylamine, 2-1,4-thiazaperhydroin-4-yl-4-pyridyl methylamine
PubChem CID 18525849
IUPAC Name (2-thiomorpholin-4-ylpyridin-4-yl)methanamine
SMILES C1CSCCN1C2=NC=CC(=C2)CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 209.32
Percent Purity 97%
Chemical Name or Material (2-Thiomorpholinopyrid-4-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.