missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenecarboxamide, 99%, Thermo Scientific™

Product Code. 10412841 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10412841 5 g Glass bottle 5g
10617874 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10412841 Supplier Thermo Scientific Acros Supplier No. 305420050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5813-89-8
Molecular Formula C5H5NOS
Molecular Weight (g/mol) 127.17
MDL Number MFCD00010425
InChI Key DENPQNAWGQXKCU-UHFFFAOYSA-N
Synonym 2-thiophenecarboxamide, 2-thienylamide, 2-thenamide, thiophen-2-carboxamide, 2-aminocarbonyl thiophene, 2-thienylamide french, thiophenecarboxamide, thiolamide, acmc-1ao1m, 5-18-06-00162 beilstein handbook reference
PubChem CID 22063
IUPAC Name thiophene-2-carboxamide
SMILES C1=CSC(=C1)C(=O)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 179.0°C to 182.0°C
Color Gray to White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 127.17
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Thiophenecarboxamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.