missing translation for 'onlineSavingsMsg'
Learn More

2-Thiophenecarboxylic acid hydrazide, 97%

Product Code. 10645572 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packaging Pakningsstørrelse
10645572 25 g Glass bottle 25g
10406960 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 10645572 Leverandør Thermo Scientific Acros Leverandørnr. 138850250
 Restricted Item
Estimated Shipment: 27-11-2026
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 2361-27-5 | C5H6N2OS | 142.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 2361-27-5
Molecular Formula C5H6N2OS
Molecular Weight (g/mol) 142.18
MDL Number MFCD00005435
InChI Key SOGBOGBTIKMGFS-UHFFFAOYSA-N
Synonym 2-thiophenecarboxylic acid hydrazide, thiophene-2-carboxylic acid hydrazide, 2-thenoylhydrazine, 2-thiophenecarboxylic hydrazide, 2-thiophenecarbohydrazide, thiophene-2-carboxylic hydrazide, 2-thiophenecarboxylic acid, hydrazide, 2-thiophenecarbohydrazonic acid, 2-thienylcarboxylic acid hydrazide, 2-thiophene carboxylic hydrazide
PubChem CID 73098
IUPAC Name thiophene-2-carbohydrazide
SMILES C1=CSC(=C1)C(=O)NN

Tekniske data

CAS Min % 96.0
CAS Max % 100.0
Melting Point 136.0°C to 139.0°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Beilstein 18, 291
Formula Weight 142.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Thiophenecarboxylic acid hydrazide, 97%
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • 2-Thiophenecarboxylic acid hydrazide
Signalord
  • Warning
Farekategori
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.