missing translation for 'onlineSavingsMsg'
Learn More

2-Thiouracil, 99%, Thermo Scientific Chemicals

Product Code. 10725821 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10725821 5 g Glass bottle 5g
10103060 100 g Plastic bottle 100g
10561184 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10725821 Supplier Thermo Scientific Acros Supplier No. 151840050
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

2-Thiouracil, Thiouracil or 4-Hydroxy-2-mercaptopyrimidine, CAS # 141-90-2, is the modified analog of the nucleotide base, uracil.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description


• 2-Thiouracil (Thiouracil or 4-Hydroxy-2-mercaptopyrimidine) is a nucleotide analog with a C-2 thioxo substitution
• 2-Thiouracil is a melanocyte suppressor and thyroid inhibitor


Applications
• 2-Thiouracil can be used to inhibit nitric oxide synthase and suppresses nitric oxide signaling in vitro
• 2-Thiouracil can be used for labeling of RNA in vitro
• 2-Thiouracil is intended for research applications only

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-90-2
Molecular Formula C4H4N2OS
Molecular Weight (g/mol) 128.15
MDL Number MFCD00006039
InChI Key ZEMGGZBWXRYJHK-UHFFFAOYSA-N
Synonym 2-thiouracil, thiouracil, 4-hydroxy-2-mercaptopyrimidine, antagothyroid, antagothyroil, deracil, nobilen, 2-thioxo-2,3-dihydropyrimidin-4 1h-one, 2-mercaptopyrimidin-4-ol, 2-mercapto-4-hydroxypyrimidine
PubChem CID 1269845
ChEBI CHEBI:348530
IUPAC Name 2-sulfanylidene-1H-pyrimidin-4-one
SMILES O=C1NC(=S)NC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point >300.0°C
Color White to Cream
Loss on Drying 0.5% max. (105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 24, 323
Merck Index 15, 9520
Solubility Information Solubility in water: 0.5g/L. Other solubilities: soluble in alkaline solutions
Formula Weight 128.15
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Thiouracil
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.