missing translation for 'onlineSavingsMsg'
Learn More

2-Thiouracil, TRC

Product Code. 30134024
Change view
Click to view available options
Quantity:
50 mg
1 g
100 g
Unit Size:
100g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134024 50 mg 50mg
30137105 100 g 100g
30005401 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134024 Supplier Toronto Research Chemicals Supplier No. TRCT37540050MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-90-2
Molecular Formula C4 H4 N2 O S
Molecular Weight (g/mol) 128.15
Synonym 4(1H)-Pyrimidinone, 2,3-dihydro-2-thioxo-, Uracil, 2-thio- (8CI), 2-Mercapto-4-hydroxypyrimidine, 2-Mercapto-4-pyrimidinol, 2-Mercapto-4-pyrimidinone, 2-Thiouracil, 2-Thioxo-2,3-dihydropyrimidin-4(1H)-one, 4-Hydroxy-2-mercaptopyrimidine, 4-Hydroxy-2-pyrimidinethiol, 6-Hydroxy-2-mercaptopyrimidine, Antagothyroil, Deracil, NSC 19473, NSC 290412, NSC 290413, NSC 290414, Nobilen, Thiouracil
IUPAC Name 2-sulfanylidene-1H-pyrimidin-4-one
SMILES O=C1NC(=S)NC=C1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C4H4N2OS/c7-3-1-2-5-4(8)6-3/h1-2H,(H2,5,6,7,8)
Formula Weight 128.0044
Percent Purity >95%
Chemical Name or Material 2-Thiouracil
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.