missing translation for 'onlineSavingsMsg'
Learn More

2-Thioxanthine, 98+%

Product Code. 11467224
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11467224 5 g 5g
11457224 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11467224 Supplier Thermo Scientific Alfa Aesar Supplier No. B21606.06
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2487-40-3 | C5H4N4OS | 168.174 g/mol

2-Thioxanthine used to inhibit MPO (Myeloperoxidase).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Thioxanthine used to inhibit MPO (Myeloperoxidase).

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2487-40-3
Molecular Formula C5H4N4OS
Molecular Weight (g/mol) 168.174
MDL Number MFCD00031505
InChI Key XNHFAGRBSMMFKL-UHFFFAOYSA-N
Synonym 2-thioxanthine, 6-hydroxy-2-mercaptopurine, 2-mercapto-9h-purin-6-ol, 2-thioxanthene, 2-mercapto-6-hydroxypurine, 2-thio-6-oxypurine, 6-hydroxypurine-2-thiol, 2-thioxo-2,3-dihydro-1h-purin-6 7h-one, xanthine, 2-thio, 6h-purin-6-one, 1,2,3,7-tetrahydro-2-thioxo
PubChem CID 1268185
IUPAC Name 2-sulfanylidene-3,7-dihydropurin-6-one
SMILES C1=NC2=C(N1)C(=O)NC(=S)N2

Specifications

Melting Point >300°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 168.17
Percent Purity ≥98%
Chemical Name or Material 2-Thioxanthine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.