missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Toluidine, TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | o-Toluidine |
| CAS | 95-53-4 |
| Molecular Formula | C7 H9 N |
| Quantity | 100 g |
| Synonym | 2-Methylbenzenamine, Prilocaine Hydrochloride Imp. B (EP), o-Toluidine, Benzenamine','2-methyl- (9CI','ACI), 2-Methylbenzenamine (ACI), o-Toluidine (8CI), 1-Amino-2-methylbenzene, 2-Amino-1-methylbenzene, 2-Aminotoluene, 2-Methyl-1-aminobenzene, 2-Methylaniline, 2-Methylphenylamine, 2-Toluidine, 2-Tolylamine, 6-Methylaniline, NSC 15348, o-Aminotoluene, o-Methylaniline, o-Methylbenzenamine, o-Tolylamine |
| InChI Formula | InChI=1S/C7H9N/c1-6-4-2-3-5-7(6)8/h2-5H,8H2,1H3 |
| SMILES | Cc1ccccc1N |
| IUPAC Name | 2-methylaniline |
| Molecular Weight (g/mol) | 107.15 |
| Formula Weight | 107.0735 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction