missing translation for 'onlineSavingsMsg'
Learn More

2-Tolylboronic acid, 95%

Product Code. 10643363 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10643363 5 g Glass bottle 5g
10471111 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10643363 Supplier Thermo Scientific Acros Supplier No. 309260050
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 16419-60-6 | C7H9BO2 | 135.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16419-60-6
Molecular Formula C7H9BO2
Molecular Weight (g/mol) 135.96
MDL Number MFCD00093526
InChI Key NSJVYHOPHZMZPN-UHFFFAOYSA-N
Synonym 2-tolylboronic acid, o-tolylboronic acid, 2-methylbenzeneboronic acid, 2-methylphenyl boronic acid, o-methylphenylboronic acid, 2-methylphenyl boranediol, tolylboronic acid, o-tolueneboronic acid, toluene-2-boronic acid, boronic acid, methylphenyl
PubChem CID 2733267
IUPAC Name (2-methylphenyl)boronic acid
SMILES CC1=CC=CC=C1B(O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 164.0°C to 165.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3C6H4B(OH)2
Quantity 5 g
Formula Weight 135.96
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-Tolylboronic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.