missing translation for 'onlineSavingsMsg'
Learn More

2-(Tri-n-butylstannyl)benzo[b]thiophene, 95%, Thermo Scientific™

Product Code. 15421578
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15421578 250 mg 250mg
15401578 1 g 1g
15411578 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15421578 Supplier Thermo Scientific Alfa Aesar Supplier No. H55348.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148961-88-0
Molecular Formula C20H32SSn
Molecular Weight (g/mol) 423.246
MDL Number MFCD01318936
InChI Key UJPVMOUQRNYNSA-UHFFFAOYSA-N
Synonym 2-tributylstannylbenzo b thiophene, benzo b thiophen-2-yltributylstannane, stannane,benzo b thien-2-yltributyl, 2-tri-n-butylstannyl benzo b thiophene, acmc-20anmg, 2-benzothienyl tributyltin, benzo b thien-2-yltributylstannane, benzo b thien-2-yltributyl-stannane, 1-benzothien-2-yl tributyl stannane, benzo b thiophen-2-yl-tributyltin
PubChem CID 2735461
IUPAC Name 1-benzothiophen-2-yl(tributyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC2=CC=CC=C2S1

Specifications

Color Colorless
Density 1.184 g/mL
Refractive Index 1.564
Quantity 250 mg
UN Number UN2788
Formula Weight 423.22
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-(Tri-n-butylstannyl)benzo[b]thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.