missing translation for 'onlineSavingsMsg'
Learn More

2-(Tri-n-butylstannyl)pyridine, tech. 80%

Product Code. 11491387
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491387 5 g 5g
11481387 1 g 1g
11401397 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491387 Supplier Thermo Scientific Alfa Aesar Supplier No. B24911.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 17997-47-6 | C17H31NSn | 368.152 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 17997-47-6
Molecular Formula C17H31NSn
Molecular Weight (g/mol) 368.152
MDL Number MFCD00052052
InChI Key GYUURHMITDQTRU-UHFFFAOYSA-N
Synonym 2-tributylstannyl pyridine, 2-tri-n-butylstannylpyridine, 2-tributylstannylpyridine, tributyl 2-pyridyl tin, 2-pyridyltributyltin, 2-1,1,1-tributylstannyl pyridine, 2-pyridyltri-n-butyltin, 2-tributylstannyl-pyridine, tri-n-butyl 2-pyridyl tin
PubChem CID 607784
IUPAC Name tributyl(pyridin-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CC=N1

Tekniske data

Density 1.15
Boiling Point 130°C to 132°C (0.8 mmHg)
Refractive Index 1.513
Quantity 5 g
UN Number UN2788
Beilstein 479329
Formula Weight 368.15
Percent Purity 80%
Grade Technical
Chemical Name or Material 2-(Tri-n-butylstannyl)pyridine

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.