missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoroacetyl)thiophene, 98%, Thermo Scientific™

Product Code. p-4228928 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10585290 1 g 1g
10854161 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10585290 Supplier Thermo Scientific Acros Supplier No. 434700010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 651-70-7
Molecular Formula C6H3F3OS
Molecular Weight (g/mol) 180.15
InChI Key CZYKJGCKVBXLGF-UHFFFAOYSA-N
Synonym 2-trifluoroacetyl thiophene, 2,2,2-trifluoro-1-thiophen-2-yl ethanone, ethanone, 2,2,2-trifluoro-1-2-thienyl, 2,2,2-trifluoro-1-2-thienyl ethanone, ethanone,2,2,2-trifluoro-1-2-thienyl, 2,2,2-trifluoro-1-thiophen-2-yl ethan-1-one, 2,2,2-trifluoro-1-2-thienyl ethan-1-one, acmc-20aolw, trifluoroacetylthiophene, 2-trifluoroacetylthiophene
PubChem CID 136464
IUPAC Name 2,2,2-trifluoro-1-thiophen-2-ylethanone
SMILES C1=CSC(=C1)C(=O)C(F)(F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 163.0°C to 164.0°C
Flash Point 59°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 180.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-(Trifluoroacetyl)thiophene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-(Trifluoroacetyl) thiophene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.