missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethoxy)benzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 11473474
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473474 1 g 1g
11483474 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11473474 Supplier Thermo Scientific Alfa Aesar Supplier No. A10707.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103008-51-1
Molecular Formula C7H4ClF3O3S
Molecular Weight (g/mol) 260.61
MDL Number MFCD00052316
InChI Key JLTBWYIUMFEOTN-UHFFFAOYSA-N
Synonym 2-trifluoromethoxy benzenesulfonyl chloride, 2-trifluoromethoxy benzene-1-sulfonyl chloride, 2-trifluoromethoxy benzenesulphonyl chloride, 2-trifluoromethoxy-benzenesulfonyl chloride, 2-trifluoromethoxy benzenesulfonylchloride, 2-trifluoromethoxy phenylsulfonyl chloride, benzenesulfonyl chloride, 2-trifluoromethoxy, trifluoromethoxyphenylsulfonyl chloride, pubchem5184, 2-trifluoromethoxyphenyl-sulfonylchloride
PubChem CID 2777215
IUPAC Name 2-(trifluoromethoxy)benzenesulfonyl chloride
SMILES FC(F)(F)OC1=CC=CC=C1S(Cl)(=O)=O

Specifications

Melting Point 29°C to 32°C
Boiling Point 75°C to 77°C (1 mmHg)
Refractive Index 1.4825
Quantity 1 g
UN Number UN3261
Beilstein 8482182
Sensitivity Moisture sensitive
Formula Weight 260.62
Percent Purity 97%
Chemical Name or Material 2- (Trifluoromethoxy)benzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.