missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)acrylic acid, 98%

Product Code. 11410597
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410597 1 g 1g
11420597 5 g 5g
11430597 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410597 Supplier Thermo Scientific Alfa Aesar Supplier No. B22443.03
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 381-98-6 | C4H2F3O2 | 139.05 g/mol

2-(Trifluoromethyl)acrylic acid is used as an acidic functional monomer for molecular imprinting of nicotine. And also used in the conventional functional monomer methacrylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trifluoromethyl)acrylic acid is used as an acidic functional monomer for molecular imprinting of nicotine. And also used in the conventional functional monomer methacrylic acid.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 381-98-6
Molecular Formula C4H2F3O2
Molecular Weight (g/mol) 139.05
MDL Number MFCD00042424
InChI Key VLSRKCIBHNJFHA-UHFFFAOYSA-M
Synonym 2-trifluoromethyl acrylic acid, 2-trifluoromethyl propenoic acid, 2-trifluoromethyl prop-2-enoic acid, tfmaa, trifluoromethacrylic acid, trifluoromethylacrylic acid, 2-trifluoromethylacrylic acid, 2-trifluoromethyl-acrylic acid, 2-propenoic acid, 2-trifluoromethyl, pubchem22654
PubChem CID 587694
SMILES [O-]C(=O)C(=C)C(F)(F)F

Specifications

Melting Point 49°C to 51°C
Flash Point 82°C (180°F)
Quantity 1 g
UN Number UN3261
Solubility Information Soluble in water.
IUPAC Name 2-(trifluoromethyl)prop-2-enoic acid
Formula Weight 140.06
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethyl)acrylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.