missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)phenothiazine, 98%, Thermo Scientific™

Product Code. 11479956
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11479956 25 g 25g
11469956 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479956 Supplier Thermo Scientific Alfa Aesar Supplier No. B20919.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-30-8
Molecular Formula C13H8F3NS
Molecular Weight (g/mol) 267.269
MDL Number MFCD00005018
InChI Key RKGYJVASTMCSHZ-UHFFFAOYSA-N
Synonym 2-trifluoromethyl phenothiazine, 2-trifluoromethyl-10h-phenothiazine, trifluoromethylphenothiazine, 2-trifluoromethylphenothiazine, 2-trifluoromethyl-phenothiazine, 10h-phenothiazine, 2-trifluoromethyl, phenothiazine, 2-trifluoromethyl, 2-triflouomethyl phenothiazine, acmc-209rf8, 4-27-00-01352 beilstein handbook reference
PubChem CID 7082
IUPAC Name 2-(trifluoromethyl)-10H-phenothiazine
SMILES C1=CC=C2C(=C1)NC3=C(S2)C=CC(=C3)C(F)(F)F

Specifications

Melting Point 188°C to 190°C
Quantity 25 g
Beilstein 226580
Formula Weight 267.27
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethyl)phenothiazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.