missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-(Trifluoromethyl)piperazine, 97%, Thermo Scientific Chemicals

Product Code. 15415257
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15415257 250 mg 250mg
15405257 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15415257 Supplier Thermo Scientific Alfa Aesar Supplier No. H32524.MD
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131922-05-9
Molecular Formula C5H9F3N2
Molecular Weight (g/mol) 154.136
MDL Number MFCD07373382
InChI Key YNEJOOCZWXGXDU-UHFFFAOYSA-N
Synonym 2-trifluoromethyl piperazine, 2-trifluoromethyl-piperazine, +/--2-trifluoromethyl piperazine, piperazine, 2-trifluoromethyl, -2-trifluoromethyl piperazine, piperazine, 2-trifluoromethyl-9ci, acmc-20aolo, 2trifluoromethylpiperazine, m-trifluoromethylpiperazine
PubChem CID 18519725
IUPAC Name 2-(trifluoromethyl)piperazine
SMILES C1CNC(CN1)C(F)(F)F

Specifications

Melting Point 94°C to 97°C
Quantity 250 mg
UN Number UN3259
Sensitivity Air Sensitive
Formula Weight 154.13
Percent Purity 97%
Chemical Name or Material (+/-)-2-(Trifluoromethyl)piperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.